Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Neuraminidase from
Clostridium perfringens (C. welchii)
Synonym(s):
Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase
CAS Number:
9001-67-6
EC Number:
232-624-6
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
N2876
Neuraminidase from
Clostridium perfringens (C. welchii)
,
Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Expand
View Pricing
N2133
Neuraminidase from
Clostridium perfringens (C. welchii)
,
Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Expand
View Pricing
N3001
Neuraminidase from
Clostridium perfringens (C. welchii)
,
Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Expand
View Pricing
N5631
Neuraminidase from
Clostridium perfringens (C. welchii)
,
Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Expand
View Pricing