Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
vacuum filtration system
Products
Product Category
filters (11)
Brand
Sigma-Aldrich (8)
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Filter Capacity (ml)
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z358193
Nalgene
®
vacuum filtration system,
filter capacity 500 mL, pore size 0.2 μm
Expand
View Pricing
Z358207
Nalgene
®
vacuum filtration system,
filter capacity 1000 mL, pore size 0.2 μm
Expand
View Pricing
Z370606
Nalgene
®
vacuum filtration system,
filter capacity 250 mL, pore size 0.2 μm
Expand
View Pricing
Z377503
Nalgene
®
vacuum filtration system,
pore size 0.45 μm
Expand
View Pricing
Z370622
Nalgene
®
vacuum filtration system,
filter capacity 500 mL, pore size 0.45 μm
Expand
View Pricing
Z358185
Nalgene
®
vacuum filtration system,
filter capacity 150 mL, pore size 0.2 μm
Expand
View Pricing
Z370657
Nalgene
®
vacuum filtration system,
capacity 1,000 mL, pore size 0.45 μm
Expand
View Pricing
Z377481
Nalgene
®
vacuum filtration system,
filter capacity 115 mL, pore size 0.2 μm
Expand
View Pricing
Z370649
Nalgene
®
vacuum filtration system,
filter capacity 500 mL, pore size 0.45 μm
Expand
View Pricing
Z370614
Nalgene
®
vacuum filtration system,
filter capacity 250 mL, pore size 0.45 μm
Expand
View Pricing
Z370592
Nalgene
®
vacuum filtration system,
filter capacity 150 mL, pore size 0.45 μm
Expand
View Pricing
TMO566-0010
Nalgene
®
vacuum filtration system,
filter capacity 500 mL, polyethersulfone, pore size 0.1 μm, case of 12 ea
Expand
View Pricing
TMO569-0020
Nalgene
®
vacuum filtration system,
filter capacity 500 mL, polyethersulfone, pore size 0.2 μm, case of 12 ea
Expand
View Pricing
TMO565-0010
Nalgene
®
vacuum filtration system,
filter capacity 150 mL, polyethersulfone, pore size 0.1 μm, case of 12 ea
Expand
View Pricing
TMO567-0010
Nalgene
®
vacuum filtration system,
filter capacity 1000 mL, polyethersulfone, pore size 0.1 μm, case of 12 ea
Expand
View Pricing
TMO568-0010
Nalgene
®
vacuum filtration system,
filter capacity 250 mL, polyethersulfone, pore size 0.1 μm, case of 12 ea
Expand
View Pricing