Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
square graduated bottle
Products
Product Category
bottles (4)
Manufacturer
Capacity (mL)
Color
Package Size
Special Grade
Natural (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
TMO312018-0125
Nalgene
®
square graduated bottle,
high-density polyethylene, bottle capacity 125 mL, natural, case of 250 ea
Expand
View Pricing
TMO312018-1000
Nalgene
®
square graduated bottle,
high-density polyethylene, bottle capacity 1000 mL, natural, case of 250 ea
Expand
View Pricing
TMO312018-0250
Nalgene
®
square graduated bottle,
high-density polyethylene, bottle capacity 250 mL, natural, case of 250 ea
Expand
View Pricing
TMO312018-0500
Nalgene
®
square graduated bottle,
high-density polyethylene, bottle capacity 500 mL, natural, case of 250 ea
Expand
View Pricing