Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
cryogenic vials
Products
Product Category
vials (7)
Brand
Sigma-Aldrich (7)
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
V5007
Nalgene
®
cryogenic vials,
capacity 2.0 mL, sterile, External threads
Expand
View Pricing
V5257
Nalgene
®
cryogenic vials,
capacity 5 mL, sterile
Expand
View Pricing
V5132
Nalgene
®
cryogenic vials,
capacity 2.0 mL, sterile, External threads, pkg of (Bulk packed)
Expand
View Pricing
V4757
Nalgene
®
cryogenic vials,
capacity 1.2 mL, External threads, sterile; irradiated
Expand
View Pricing
V4632
Nalgene
®
cryogenic vials,
capacity 2.0 mL, non-sterile
Expand
View Pricing
V4882
Nalgene
®
cryogenic vials,
capacity 1.2 mL, sterile; irradiated, External threads, pkg of (Bulk packed)
Expand
View Pricing
V4507
Nalgene
®
cryogenic vials,
capacity 1.2 mL, non-sterile, External threads
Expand
View Pricing