Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Transfer Pipette
Products
Product Category
pipettes (24)
Manufacturer
Length (mm)
Package Size
Sterility
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
MTCP412111
MTC
™
Bio Transfer Pipette,
size 1.5 mL, small bulb, extended tip, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP412211
MTC
™
Bio Transfer Pipette,
size 5 mL, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP411100
MTC
™
Bio Transfer Pipette,
size 5 mL, large bulb, non-sterile, pack of 250 ea (bulk bag)
Expand
View Pricing
MTCP411711
MTC
™
Bio Transfer Pipette,
size 3 mL, short bulb, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP411700
MTC
™
Bio Transfer Pipette,
size 3 mL, short bulk, non-sterile, pack of 250 ea (bulk bag)
Expand
View Pricing
MTCP411511
MTC
™
Bio Transfer Pipette,
size 5 mL, blood bank, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP411500
MTC
™
Bio Transfer Pipette,
size 5 mL, blood bank, non-sterile, pack of 250 ea (bulk bag)
Expand
View Pricing
MTCP413311
MTC
™
Bio Transfer Pipette,
size 4 mL, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP412101
MTC
™
Bio Transfer Pipette,
size 1.5 mL, small bulb, extended tip, non-sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP412301
MTC
™
Bio Transfer Pipette,
size 7 mL, large bulb, non-sterile, pack of 400 ea (bulk bag)
Expand
View Pricing
MTCP411311
MTC
™
Bio Transfer Pipette,
size 5 mL, large bulb, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP411400
MTC
™
Bio Transfer Pipette,
size 7 mL, large bulb, non-sterile, pack of 250 ea (bulk bag)
Expand
View Pricing
MTCP411411
MTC
™
Bio Transfer Pipette,
size 7 mL, large bulb, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP411414
MTC
™
Bio Transfer Pipette,
size 7 mL, large bulb, sterile, pack of 500 ea (525 x packs 20 ea)
Expand
View Pricing
MTCP411111
MTC
™
Bio Transfer Pipette,
size 5 mL, large bulb, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP411114
MTC
™
Bio Transfer Pipette,
size 5 mL, large bulb, sterile, pack of 25 × 500 ea (25 x packs 20 ea)
Expand
View Pricing
MTCP412201
MTC
™
Bio Transfer Pipette,
size 5 mL, extended tip, non-sterile, pack of 500 ea (bulk bag)
Expand
View Pricing
MTCP412311
MTC
™
Bio Transfer Pipette,
size 7 mL, large bulb, extended tip, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCP413200
MTC
™
Bio Transfer Pipette,
size 4 mL, short stem, non-sterile, pack of 250 ea (bulk bag)
Expand
View Pricing
MTCP413211
MTC
™
Bio Transfer Pipette,
size 4 mL, short stem, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
Page 1 of 2
Page 1 of 2