Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Fraction CollectorTube Rack
Products
Product Category
racks (6)
Manufacturer
Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
MTCR1040
MTC
™
Bio Fraction CollectorTube Rack,
holds 80 x 1.5 /2.0 mL tubes, assorted colors, pack of 5 ea
Expand
View Pricing
MTCR1040Y
MTC
™
Bio Fraction CollectorTube Rack,
holds 80 x 1.5 /2.0 mL tubes, yellow, pack of 5 ea
Expand
View Pricing
MTCR1040G
MTC
™
Bio Fraction CollectorTube Rack,
holds 80 x 1.5 /2.0 mL tubes, green, pack of 5 ea
Expand
View Pricing
MTCR1040B
MTC
™
Bio Fraction CollectorTube Rack,
holds 80 x 1.5 /2.0 mL tubes, blue, pack of 5 ea
Expand
View Pricing
MTCR1040R
MTC
™
Bio Fraction CollectorTube Rack,
holds 80 x 1.5 /2.0 mL tubes, red, pack of 5 ea
Expand
View Pricing
MTCR1040O
MTC
™
Bio Fraction CollectorTube Rack,
holds 80 x 1.5 /2.0 mL tubes, orange, pack of 5 ea
Expand
View Pricing