Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Monel
®
alloy 400
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
GF45995158
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, size 100 × 100 mm, temper annealed
Expand
View Pricing
GF96661637
Monel
®
alloy 400,
foil, Ni65%/Cu33%/Fe 2%, thickness 0.3 mm, length 2 m, temper annealed
Expand
View Pricing
GF32352735
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, 10 mm diameter, temper annealed
Expand
View Pricing
GF32959334
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, size 25 × 25 mm, temper annealed
Expand
View Pricing
GF30954337
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, 25 mm diameter, temper annealed
Expand
View Pricing
GF35440747
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, 8 mm diameter, temper annealed
Expand
View Pricing
GF94171978
Monel
®
alloy 400,
foil, thickness 0.3 mm, Ni65%/Cu33%/Fe 2%, size 50 × 50 mm, temper annealed
Expand
View Pricing
GF26024622
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, 50 mm diameter, temper annealed
Expand
View Pricing
GF34173884
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, 15 mm diameter, temper annealed
Expand
View Pricing
GF91737456
Monel
®
alloy 400,
foil, thickness 0.3 mm, Ni65%/Cu33%/Fe 2%, length 1 m, temper annealed
Expand
View Pricing
GF26355618
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, 6 mm diameter, temper annealed
Expand
View Pricing
GF28319654
Monel
®
alloy 400,
foil, Ni 65%/Cu 33%/Fe 2, thickness 0.3 mm, length 0.5 m, temper annealed
Expand
View Pricing
GF62203890
Monel
®
alloy 400,
foil, Ni65%/Cu33%/Fe 2%, thickness 0.3 mm, size 200 × 200 mm, temper annealed
Expand
View Pricing
GF97050720
Monel
®
alloy 400,
foil, Ni65%/Cu33%/Fe 2%, thickness 0.3 mm, diameter 4 mm, temper annealed
Expand
View Pricing