Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molecular sieves, 13X
Linear Formula:
Na
86
[AlO
2
)
86
(SiO
2
)
106
] · xH
2
O
CAS Number:
63231-69-6
Products
Product Category
sieves (5)
Brand
Sigma-Aldrich (5)
Physical Form
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
283592
Molecular sieves, 13X,
powder, ~2 μm avg. part. size
Expand
View Pricing
208639
Molecular sieves, 13X,
beads, 4-8 mesh
Expand
View Pricing
208647
Molecular sieves, 13X,
beads, 8-12 mesh
Expand
View Pricing
334340
Molecular sieves, 13X,
pellets, 1.6 mm diameter
Expand
View Pricing
334359
Molecular sieves, 13X,
pellets, 3.2 mm
Expand
View Pricing