Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microtube Rack
Products
Product Category
racks (10)
Manufacturer
Diameter (mm)
Height (mm)
Length (mm)
Color
Package Size
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
HS29025G
Microtube Rack,
assorted colors rack
Expand
View Pricing
HS29025E
Microtube Rack,
yellow
Expand
View Pricing
HS120243
Microtube Rack,
blue Nylon, pack of 4 racks, Holds 96 x 1.5-2 mL tubes
Expand
View Pricing
HS29025F
Microtube Rack,
orange rack
Expand
View Pricing
HS29025C
Microtube Rack,
green
Expand
View Pricing
HS120241
Microtube Rack,
blue Nylon, pack of 8 racks, Holds 24 x 1.5-2 mL tubes
Expand
View Pricing
HS29025A
Microtube Rack,
natural
Expand
View Pricing
HS29025J
Microtube Rack,
red
Expand
View Pricing
HS29025B
Microtube Rack,
blue
Expand
View Pricing
HS29025D
Microtube Rack,
pink rack
Expand
View Pricing