Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microtube Centrifuge Tubes
Products
Product Category
tubes (3)
Brand
Sigma-Aldrich (3)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BMSC1005SC5S
Microtube Centrifuge Tubes,
capacity 5 mL, clear tube, sterile, cap (screw, attached), pack of 500 ea (10 x racks 50ea)
Expand
View Pricing
BMSC1005SC5OB
Microtube Centrifuge Tubes,
capacity 5 mL, black tube, sterile, cap (screw, attached), pack of 500 ea
Expand
View Pricing
BMSC1005SC5
Microtube Centrifuge Tubes,
capacity 5 mL, clear tube, non-sterile, cap (screw, separate), pack of 500 ea (5 x bags 100ea)
Expand
View Pricing