Skip to Content
Merck
Methyl 3,5-dihydroxyphenylacetate 97%

Methyl 3,5-dihydroxyphenylacetate

Methyl 3,5-dihydroxyphenylacetate 97%
Linear Formula:
(HO)2C6H3CH2CO2CH3
CAS Number:
Molecular Weight:
182.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing