Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methacrylic acid
Synonym(s):
2-Methacrylic acid, 2-Methylpropenoic acid, Methacrylic acid
Linear Formula:
H
2
C=C(CH
3
)COOH
CAS Number:
79-41-4
Molecular Weight:
86.09
EC Number:
201-204-4
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
155721
Methacrylic acid,
contains 250 ppm MEHQ as inhibitor, 99%
Expand
View Pricing
8.00578
Methacrylic acid,
(stabilised with hydroquinone monomethyl ether) for synthesis
Expand
View Pricing
19-2110
Methacrylic acid,
SAJ first grade, ≥98.0%
Expand
View Pricing