Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mercury(II) chloride
Synonym(s):
Mercuric chloride, Mercury(II) chloride
Linear Formula:
HgCl
2
CAS Number:
7487-94-7
Molecular Weight:
271.50
EC Number:
231-299-8
Products
Product Category
salts (6)
Brand
Sigma-Aldrich (3)
Supelco (4)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
215465
Mercury(II) chloride,
ACS reagent, ≥99.5%
Expand
View Pricing
1.04419
Mercury(II) chloride,
for analysis EMSURE
®
Reag. Ph Eur,ACS
Expand
View Pricing
M1136
Mercury(II) chloride,
ReagentPlus
®
, 99%
Expand
View Pricing
1.04417
Mercury(II) chloride,
fine cryst. EMPLURA
®
Expand
View Pricing
1.93203
Mercury(II) chloride,
for analysis EMPARTA
®
ACS
Expand
View Pricing
1.93603
Mercury(II) chloride,
EMPLURA
®
Expand
View Pricing
413445
Mercury(II) chloride,
≥98%
Expand
View Pricing