Mebendazole
Synonym(s):
5-Benzoyl-2-benzimidazolecarbamic acid methyl ester, Mebendazol, Methyl N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate
Empirical Formula (Hill Notation):
C16H13N3O3
CAS Number:
Molecular Weight:
295.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing