![Manganese(II) chloride AnhydroBeads™, −10 mesh, 99.999% trace metals basis](/deepweb/assets/sigmaaldrich/product/structures/122/343/37d3c335-e720-49a9-b3f9-8fc6bc199241/640/37d3c335-e720-49a9-b3f9-8fc6bc199241.png)
Manganese(II) chloride
![Manganese(II) chloride AnhydroBeads™, −10 mesh, 99.999% trace metals basis](/deepweb/assets/sigmaaldrich/product/structures/122/343/37d3c335-e720-49a9-b3f9-8fc6bc199241/640/37d3c335-e720-49a9-b3f9-8fc6bc199241.png)
Synonym(s):
Manganese dichloride, Sacchite, Manganese(II) chloride
Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing