
Lysozyme from human neutrophils

Synonym(s):
Mucopeptide N-acetylmuramoylhydrolase, Muramidase
CAS Number:
Products
Brand
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Tissue
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing