Lumefantrine
Synonym(s):
(9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-α-[(dibutylamino)methyl]-9H-fluorene-4-methanol, Benflumetol, CPG-56695, Lumefantrine
Empirical Formula (Hill Notation):
C30H32Cl3NO
CAS Number:
Molecular Weight:
528.94
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3