Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Luer-to-Tubing Connector
Synonym(s):
environmental, veterinary
Products
Product Category
connectors (10)
Brand
Sigma-Aldrich (1)
Manufacturer
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z261491
Luer-to-Tubing Connector,
bulkhead panel mounted Micro-Mate
®
female Luer to hose end for 1/16 in. to 3/32 in. I.D. tubing, nickel plated
Expand
View Pricing
Z118028
Luer-to-Tubing Connector,
male Luer lock to hose end, for tubing i.d., 1/16 ‑ 3/32 in., nickel plated
Expand
View Pricing
Z261580
Luer-to-Tubing Connector,
Micro-Mate
®
female Luer to hose end for 1/4 in. to 5/16 in. I.D. tubing, 316 stainless steel
Expand
View Pricing
Z117927
Luer-to-Tubing Connector,
female Luer lock to hose end, for tubing i.d., 1/8 ‑ 3/16 in., nickel plated
Expand
View Pricing
Z261599
Luer-to-Tubing Connector,
Micro-Mate
®
female Luer to hose end for 1/8 in. to 3/16 in. I.D. tubing, 316 stainless steel
Expand
View Pricing
Z261483
Luer-to-Tubing Connector,
bulkhead panel mounted Micro-Mate
®
female Luer to hose end for 1/8 in. to 3/16 in. I.D. tubing, nickel plated
Expand
View Pricing
Z261475
Luer-to-Tubing Connector,
Micro-Mate
®
female Luer to hose end for 1/16 in. to 3/32 in. I.D. Tubing, nickel plated
Expand
View Pricing
Z117935
Luer-to-Tubing Connector,
female Luer to hose end, for tubing i.d., 1/16 ‑ 3/32 in., nickel plated
Expand
View Pricing
Z261610
Luer-to-Tubing Connector,
bulkhead panel mounted Micro-Mate
®
female Luer to hose end for 1/8 in. to 3/16 in. I.D. tubing, 316 stainless steel
Expand
View Pricing
Z261629
Luer-to-Tubing Connector,
bulkhead panel mounted Micro-Mate
®
female Luer to hose end for 1/16 in. to 3/32 in. I.D. tubing, 316 stainless steel
Expand
View Pricing