Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Ornithine monohydrochloride
Synonym(s):
(
S
)-2,5-Diaminopentanoic acid monohydrochloride, L-Ornithine monohydrochloride
Empirical Formula (Hill Notation):
C
5
H
12
N
2
O
2
· HCl
CAS Number:
3184-13-2
Molecular Weight:
168.62
EC Number:
221-678-6
Products
Product Category
amino acids (4)
attachment factors (1)
lipids (1)
stabilizers (1)
viscosity reducing agents (1)
Brand
Millipore (1)
SAFC (1)
Sigma-Aldrich (3)
Supelco (2)
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Reference Material Type
Special Grade
analytical standard (1)
certified reference material (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
O2375
L-Ornithine monohydrochloride,
≥99%
Expand
View Pricing
O6503
L-Ornithine monohydrochloride,
BioReagent, suitable for cell culture, ≥99%
Expand
View Pricing
O4386
L-Ornithine monohydrochloride,
≥99%, BioXtra
Expand
View Pricing
1.37118
L-Ornithine monohydrochloride,
EMPROVE
®
EXPERT, DAB
Expand
View Pricing
1.06906
L-Ornithine monohydrochloride,
for biochemistry
Expand
View Pricing
19943
L-Ornithine monohydrochloride,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing
57197
L-Ornithine monohydrochloride,
analytical standard
Expand
View Pricing