Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lincomycin hydrochloride
Synonym(s):
Lincocin hydrochloride, Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-2-pyrrolidinecarboxamido)-1-thio-D-
erythro
-α-D-galactooctopyranoside hydrochloride
Empirical Formula (Hill Notation):
C
18
H
34
N
2
O
6
S · HCl
CAS Number:
859-18-7
Molecular Weight:
443.00
EC Number:
212-726-7
Products
Product Category
antibiotics (4)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Optical Isomer
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
62143
Lincomycin hydrochloride,
96.0-102.0% (HPLC)
Expand
View Pricing
L6004
Lincomycin hydrochloride,
≥90% (TLC)
Expand
View Pricing
L2774
Lincomycin hydrochloride,
suitable for cell culture, BioReagent
Expand
View Pricing
BP215
Lincomycin hydrochloride,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing