Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Laser WetGrip labels
Products
Product Category
labels (4)
Color
Manufacturer
Length (in)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z743515
Laser WetGrip labels,
white, L × W 0.94 in. × 0.50 in., roll of 380
Expand
View Pricing
Z743514
Laser WetGrip labels,
white, L × W 0.875 in. × 0.875 in., roll of 2400
Expand
View Pricing
Z743516
Laser WetGrip labels,
white, L × W 1.28 in. × 0.50 in., roll of 1700
Expand
View Pricing
Z743517
Laser WetGrip labels,
white, L × W 1.50 in. × 0.75 in., roll of 1200
Expand
View Pricing