![Lamotrigine impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/322/398/99b0503c-5886-4c45-b394-9e78d209a68e/640/99b0503c-5886-4c45-b394-9e78d209a68e.png)
Lamotrigine impurity E
![Lamotrigine impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/322/398/99b0503c-5886-4c45-b394-9e78d209a68e/640/99b0503c-5886-4c45-b394-9e78d209a68e.png)
Synonym(s):
2,3-Dichlorobenzoic acid
Linear Formula:
Cl2C6H3CO2H
CAS Number:
Molecular Weight:
191.01
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing