Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Keck
®
Joint Clip
Products
Product Category
clamps (9)
Brand
Sigma-Aldrich (2)
Color
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z150428
Keck
®
Joint Clip,
standard taper, joint size 14, yellow
Expand
View Pricing
Z150436
Keck
®
Joint Clip,
standard taper, joint size 19, blue
Expand
View Pricing
Z150452
Keck
®
Joint Clip,
standard taper, joint size 29, red
Expand
View Pricing
Z222348
Keck
®
Joint Clip,
standard taper, yellow, joint size 40
Expand
View Pricing
Z222488
Keck
®
Joint Clip,
ball-joint, joint size, 35
Expand
View Pricing
Z222356
Keck
®
Joint Clip,
standard taper, joint size 45, brown
Expand
View Pricing
Z222461
Keck
®
Joint Clip,
ball-joint, joint size, 29
Expand
View Pricing
Z222313
Keck
®
Joint Clip,
standard taper, orange, joint size 34
Expand
View Pricing
Z222321
Keck
®
Joint Clip,
standard taper, violet, joint size 12
Expand
View Pricing