Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Julabo
®
CORIO
™
C open heating bath circulator
Products
Product Category
baths (9)
Manufacturer
Capacity (L)
Length (cm)
Color
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z742976
Julabo
®
CORIO
™
C open heating bath circulator,
C-BT27, capacity 20 L (27 L), transparent plastic bath, temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742979
Julabo
®
CORIO
™
C open heating bath circulator,
C-B5, capacity 3.5 L (5 L), stainless steel bath (Insulated), temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742967
Julabo
®
CORIO
™
C open heating bath circulator,
C-BT5, capacity 3.5 L (5 L), transparent plastic bath, temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742973
Julabo
®
CORIO
™
C open heating bath circulator,
C-BT19, capacity 14 L (19 L), transparent plastic bath, temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742970
Julabo
®
CORIO
™
C open heating bath circulator,
C-BT9, capacity 6 L (9 L), transparent plastic bath, temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742982
Julabo
®
CORIO
™
C open heating bath circulator,
C-B13, capacity 9 L (13 L), stainless steel bath (Insulated), temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742992
Julabo
®
CORIO
™
C open heating bath circulator,
C-B27, capacity 17 L (27 L), stainless steel bath (Insulated), temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742985
Julabo
®
CORIO
™
C open heating bath circulator,
C-B17, capacity 13 L (17 L), stainless steel bath (Insulated), temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing
Z742989
Julabo
®
CORIO
™
C open heating bath circulator,
C-B19, capacity 14 L (19 L), stainless steel bath (Insulated), temperature 20-100 °C, AC/DC input 115 V AC, US 3-pin plug (5-15P)
Expand
View Pricing