Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Jars Nalgene
®
style 2118
Products
Manufacturer
Agency
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z380261
Jars Nalgene
®
style 2118,
capacity 1000 mL
Expand
View Pricing
Z380253
Jars Nalgene
®
style 2118,
capacity 500 mL
Expand
View Pricing
Z380229
Jars Nalgene
®
style 2118,
capacity 60 mL
Expand
View Pricing
Z380245
Jars Nalgene
®
style 2118,
capacity 250 mL
Expand
View Pricing
Z380237
Jars Nalgene
®
style 2118,
capacity 125 mL
Expand
View Pricing
TMO2118-0001
Jars Nalgene
®
style 2118,
clear polypropylene copolymer, 30 mL capacity, case of 72 ea
Expand
View Pricing