Isosilybin A
Synonym(s):
(2R,3R)-3,5,7-Trihydroxy-2-[(2R,3R)-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydrobenzo[1,4]dioxin-6-yl]-4-chromanone
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing