Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Indium tin oxide coated PET
Synonym(s):
ITO-PET, Polyethylene terephthalate film, ITO coated
Linear Formula:
In
2
O
3
· (SnO
2
)
x
Products
Brand
Sigma-Aldrich (3)
Physical Form
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
639303
Indium tin oxide coated PET,
surface resistivity 60 Ω/sq, L × W × thickness 1 ft × 1 ft × 5 mil, sheet
Expand
View Pricing
749729
Indium tin oxide coated PET,
surface resistivity 60 Ω/sq, L × W × thickness 1 ft × 1 ft × 7 mil, sheet
Expand
View Pricing
749737
Indium tin oxide coated PET,
surface resistivity 100 Ω/sq, L × W × thickness 1 ft × 1 ft × 7 mil, sheet
Expand
View Pricing