Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-20 of 57
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF04344724
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
10m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF28881850
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
sheet, 100 x 100mm, 2.0mm thickness, annealed
Expand
View Pricing
GF61553022
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
rod, 200mm x 25mm diameter, annealed
Expand
View Pricing
GF87002670
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
sheet, 100 x 103mm, 5mm thickness
Expand
View Pricing
GF18793817
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
foil, 1m coil, 0.025mm thickness, annealed
Expand
View Pricing
GF30859566
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
sheet, 200 x 200mm, 2.0mm thickness, annealed
Expand
View Pricing
GF56567335
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
50m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF34986864
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
sheet, 200 x 200mm, 1.0mm thickness, annealed
Expand
View Pricing
GF53353100
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF26891411
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
rod, 500mm x 3.0mm diameter, annealed
Expand
View Pricing
GF45989207
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
rod, 500mm x 6.35mm diameter, annealed
Expand
View Pricing
GF38258264
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
foil, 150 x 150mm, 0.2mm thickness, annealed
Expand
View Pricing
GF17081029
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
rod, 50mm x 25mm diameter, annealed
Expand
View Pricing
GF09774186
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
20m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF31550318
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
rod, 1000mm x 6.35mm diameter, annealed
Expand
View Pricing
GF62613413
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
sheet, 100 x 100mm, 1.0mm thickness, annealed
Expand
View Pricing
GF01407327
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
foil, 0.5m coil, 0.10mm thickness, annealed
Expand
View Pricing
GF25597503
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
rod, 100mm x 6.35mm diameter, annealed
Expand
View Pricing
GF62792398
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
foil, 50mm disks, 0.025mm thickness, annealed
Expand
View Pricing
GF03146284
Inconel
®
625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5,
foil, 140 x 140mm, 0.025mm thickness, annealed, not light tested (nlt)
Expand
View Pricing
Page 1 of 3
Page 1 of 3