Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydrocortisone hydrogen succinate
Synonym(s):
Hydrocortisone 21-hemisuccinate, 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate, Cortisol 21-hemisuccinate
Empirical Formula (Hill Notation):
C
25
H
34
O
8
CAS Number:
2203-97-6
Molecular Weight:
462.53
EC Number:
218-612-3
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
H1425000
Hydrocortisone hydrogen succinate,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP187
Hydrocortisone hydrogen succinate,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing