Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
HisTrap
™
High Performance
Products
Manufacturer
pH Value
ID (mm)
Volume (ml)
Package Size
Particle Size (µm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GE17-5248-02
HisTrap
™
High Performance,
Cytiva 17-5248-02, pack of 5 × 5 mL
Expand
View Pricing
GE17-5247-01
HisTrap
™
High Performance,
Cytiva 17-5247-01, pack of 5 × 1 mL
Expand
View Pricing
GE17-5248-01
HisTrap
™
High Performance,
Cytiva 17-5248-01, pack of 1 × 5 mL
Expand
View Pricing
GE29-0510-21
HisTrap
™
High Performance,
Cytiva 29-0510-21, pack of 1 mL
Expand
View Pricing