Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexokinase from
Saccharomyces cerevisiae
Synonym(s):
ATP:D-Hexose-6-phosphotransferase, Hexokinase from yeast
CAS Number:
9001-51-8
EC Number:
232-611-5
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Gene Alias
Manufacturer
Physical Form
Chemical Composition
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
H4502
Hexokinase from
Saccharomyces cerevisiae
,
Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)
Expand
View Pricing
H6380
Hexokinase from
Saccharomyces cerevisiae
,
lyophilized powder, ≥350 units/mg protein, Protein ≥10 % by biuret
Expand
View Pricing