Skip to Content
Merck
Heptanes, mixture of isomers contains ≥25% n-heptane, ReagentPlus®

Heptanes, mixture of isomers

Heptanes, mixture of isomers contains ≥25% n-heptane, ReagentPlus®
Empirical Formula (Hill Notation):
C7H16
CAS Number:
Molecular Weight:
100.20

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing