Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hellma
®
fluorescence cuvettes, ultra Micro
Products
Product Category
cuvettes (6)
Manufacturer
Diameter (mm)
Volume (µL)
Detection Method
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z800090
Hellma
®
fluorescence cuvettes, ultra Micro,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 1.5x1.5 mm, chamber volume 12 μL
Expand
View Pricing
Z800201
Hellma
®
fluorescence cuvettes, ultra Micro,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 1.5x1.5 mm, chamber volume 12 μL
Expand
View Pricing
Z802336
Hellma
®
fluorescence cuvettes, ultra Micro,
High Performance Quartz Glass, Suprasil
®
quartz, spectral range 200-2500 nm, pathlength 3x3 mm, chamber volume 45 μL
Expand
View Pricing
Z802433
Hellma
®
fluorescence cuvettes, ultra Micro,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 3x3 mm, chamber volume 45 μL
Expand
View Pricing
Z802549
Hellma
®
fluorescence cuvettes, ultra Micro,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 10x2 mm, chamber volume 100 μL
Expand
View Pricing
Z802662
Hellma
®
fluorescence cuvettes, ultra Micro,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 10x2 mm, chamber volume 100 μL
Expand
View Pricing