Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner round bottom tubes, polystyrene
Products
Product Category
tubes (3)
Color
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z617865
Greiner round bottom tubes, polystyrene,
test tube, 12 mL, 16 x 100 mm, polystrene, round (U) bottom
Expand
View Pricing
Z617806
Greiner round bottom tubes, polystyrene,
test tube, 14 mL, 17 x 100 mm, polystrene, round (U) bottom
Expand
View Pricing
Z617776
Greiner round bottom tubes, polystyrene,
test tube, 20 mL, 16 x 152 mm, polystrene, round (U) bottom
Expand
View Pricing