Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner centrifuge tubes
Synonym(s):
Greiner centrifuge tubes
Products
Product Category
tubes (5)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Agency
Capacity (mL)
Diameter (mm)
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
T1943
Greiner centrifuge tubes,
centrifuge tube, 15 mL, 17x120 mm, conical (V) bottom, w/ graduation, I.D. field
Expand
View Pricing
T2318
Greiner centrifuge tubes,
centrifuge tube, 50 mL, 30 x 115 mm, conical (V) bottom, w/ graduations, I.D. field
Expand
View Pricing
T1818
Greiner centrifuge tubes,
centrifuge tube, 15 mL, 17x120 mm, conical (V) bottom, w/ graduation, I.D. field, w/ rack
Expand
View Pricing
T2193
Greiner centrifuge tubes,
capacity 50 mL, diam. × L 30 mm × 115 mm, conical bottom (with skirt)
Expand
View Pricing
Z666106
Greiner centrifuge tubes,
capacity 6.5 mL, polystyrene tube, clear, sterile
Expand
View Pricing