Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner 96 well plates, polypropylene
Products
Product Category
plates (12)
Brand
Sigma-Aldrich (2)
Color
Manufacturer
Binding Type
Wells
Working Volume (ml)
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
M8185
Greiner 96 well plates, polypropylene,
392 μL/well, V-bottom clear wells
Expand
View Pricing
M9685
Greiner 96 well plates, polypropylene,
392 μL/well, flat bottom black polypropylene wells
Expand
View Pricing
M7310
Greiner 96 well plates, polypropylene,
392 μL/well, round bottom clear wells, non-sterile
Expand
View Pricing
Z667234
Greiner 96 well plates, polypropylene,
0.5 mL/well, clear
Expand
View Pricing
M9060
Greiner 96 well plates, polypropylene,
392 μL/well, flat bottom clear polypropylene wells, non-sterile
Expand
View Pricing
M9810
Greiner 96 well plates, polypropylene,
1.2 mL, round bottom clear polypropylene wells
Expand
View Pricing
M0186
Greiner 96 well plates, polypropylene,
1.2 mL, round bottom clear polypropylene wells
Expand
View Pricing
M1561
Greiner 96 well plates, polypropylene,
2.4 mL, conical bottom clear polypropylene wells, sterile
Expand
View Pricing
Z643130
Greiner 96 well plates, polypropylene,
size 96 wells, suitable for ABI, skirt (Half skirted)
Expand
View Pricing
Z711063
Greiner 96 well plates, polypropylene,
size 96 well, skirt
Expand
View Pricing
M1186
Greiner 96 well plates, polypropylene,
2.4 mL, conical bottom clear polypropylene wells, non-sterile
Expand
View Pricing
M8935
Greiner 96 well plates, polypropylene,
392 μL/well, conical bottom black polypropylene wells, non-sterile
Expand
View Pricing