Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs wound splint, w/ suture sites
Products
Manufacturer
ID (mm)
OD (mm)
Color
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GBLRD476684
Grace Bio-Labs wound splint, w/ suture sites,
clear silicone, O.D. × I.D. × thickness 14 mm × 10 mm × 0.5 mm
Expand
View Pricing
GBLRD476687
Grace Bio-Labs wound splint, w/ suture sites,
red silicone, O.D. × I.D. × thickness 14 mm × 7 mm × 0.5 mm
Expand
View Pricing
GBLRD476685
Grace Bio-Labs wound splint, w/ suture sites,
clear silicone, O.D. × I.D. × thickness 24 mm × 20 mm × 0.5 mm
Expand
View Pricing