Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber
Products
Manufacturer
Diameter (mm)
Length (mm)
Volume (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GBL611101
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 18 μL, diam. × thickness 13 mm × 0.15 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611102
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 30 μL, diam. × thickness 20 mm × 0.15 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611104
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 30-50 μL, L × W × thickness 21.6 mm × 21.6 mm × 0.15 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611107
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 25 μL, L × W × thickness 8 mm × 20 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing
GBL612106
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 100 μL, L × W × thickness 20 mm × 21 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing
GBL612107
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 50 μL, L × W × thickness 9.8 mm × 20 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing
GBL612105
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 275-320 μL, L × W × thickness 60 mm × 21 mm × 0.15 mm, port diam. 3.2 mm
Expand
View Pricing
GBL612101
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 180-200 μL, L × W × thickness 22 mm × 40 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing
GBL612102
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 280-330 μL, L × W × thickness 22 mm × 60 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611105
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 50-100 μL, L × W × thickness 21 mm × 40 mm × 0.15 mm, port diam. 3.2 mm
Expand
View Pricing
GBL611115
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 40-70 μL, L × W × thickness 22 mm × 30 mm × 0.15 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611106
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 150-300 μL, L × W × thickness 45 mm × 45 mm × 0.15 mm, port diam. 1.5 mm
Expand
View Pricing
GBL612302
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 275-320 μL, L × W × thickness 60 mm × 21 mm × 0.25 mm, port diam. 3.0 mm
Expand
View Pricing
GBL612301
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 275-320 μL, L × W × thickness 60 mm × 21 mm × 0.25 mm, port diam. 3.2 mm, 200 port seals included
Expand
View Pricing
GBL440904
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 300 μL, L × W × thickness 21 mm × 60 mm × 0.14 mm, port diam. 3.2 mm
Expand
View Pricing
GBL612103
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 310-425 μL, L × W × thickness 22 mm × 65 mm × 0.25 mm, port diam. 3.2 mm
Expand
View Pricing
GBL611103
Grace Bio-Labs HybriWell
™
sealing system, SecureSeal
™
adhesive chamber,
chamber volume 30-50 μL, L × W × thickness 19 mm × 32 mm × 0.15 mm, port diam. 1.5 mm
Expand
View Pricing