Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber
Products
Manufacturer
Diameter (mm)
Length (mm)
Volume (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GBL612204
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber,
chamber volume 30-50 μL, L × W × thickness 22 mm × 22 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611205
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber,
chamber volume 50-100 μL, L × W × thickness 22 mm × 40 mm × 0.12 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611204
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber,
chamber volume 15-25 μL, L × W × thickness 22 mm × 22 mm × 0.12 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611202
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber,
chamber volume 140-165 μL, L × W × thickness 22 mm × 60 mm × 0.12 mm, port diam. 1.5 mm
Expand
View Pricing
GBL611201
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber,
chamber volume 90-100 μL, L × W × thickness 22 mm × 40 mm × 0.12 mm, port diam. 1.5 mm
Expand
View Pricing
GBL612202
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber,
chamber volume 280-330 μL, L × W × thickness 22 mm × 60 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing
GBL612201
Grace Bio-Labs HybriWell-FL
™
sealing system, Fluor-"friendly" adhesive chamber,
chamber volume 180-200 μL, L × W × thickness 22 mm × 40 mm × 0.25 mm, port diam. 1.5 mm
Expand
View Pricing