Skip to Content
Merck
Goserelin Acetate certified reference material, pharmaceutical secondary standard

Goserelin acetate

Goserelin Acetate certified reference material, pharmaceutical secondary standard
Synonym(s):
[D-Ser(tBu)6,Azagly10]-LHRH acetate salt, Goserelin acetate
Empirical Formula (Hill Notation):
C59H84N18O14
CAS Number:
Molecular Weight:
1269.41

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade