Glyoxylic acid solution
Synonym(s):
Formylformic acid, Oxoethanoic acid
Linear Formula:
OHCCOOH
CAS Number:
Molecular Weight:
74.04
Products
Product Category
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing