Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
γ-Globulins from bovine blood
Synonym(s):
Bovine γ-Globulin
CAS Number:
9007-83-4
EC Number:
232-706-1
Products
Product Category
proteins (2)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Purity
Tissue
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
G5009
γ-Globulins from bovine blood,
≥99% (agarose gel electrophoresis)
Expand
View Pricing
G7516
γ-Globulins from bovine blood,
≥97% (agarose gel electrophoresis)
Expand
View Pricing
SRE0011
γ-Globulins from bovine blood,
Suitable for manufacturing of diagnostic kits and reagents, ≥99% (agarose gel electrophoresis)
Expand
View Pricing