Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified
Products
Manufacturer
Physical Form
Format
Application
Reference Material Type
Filter Products
Showing 1-20 of 36
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
N1000
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 2610 mPa.s (25 °C)
Expand
View Pricing
S60
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 100.7 mPa.s (25 °C)
Expand
View Pricing
N100
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 202.6 mPa.s (25 °C)
Expand
View Pricing
S3
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 3.291 mPa.s (25 °C)
Expand
View Pricing
N35
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 55.36 mPa.s (25 °C)
Expand
View Pricing
D10
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 10.37 mPa.s (25 °C)
Expand
View Pricing
D1000
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 990.0 mPa.s (25 °C)
Expand
View Pricing
S600
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 1273 mPa.s (25 °C)
Expand
View Pricing
N10
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 14.42 mPa.s (25 °C)
Expand
View Pricing
S20
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 29.04 mPa.s (25 °C)
Expand
View Pricing
N26
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 48.59 mPa.s (25 °C)
Expand
View Pricing
N75
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 124.9 mPa.s (25 °C)
Expand
View Pricing
S6
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 7.442 mPa.s (25 °C)
Expand
View Pricing
D5000
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 5738 mPa.s (25 °C)
Expand
View Pricing
S30000
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 72328 mPa.s (25 °C)
Expand
View Pricing
D5
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 4.745 mPa.s (25 °C)
Expand
View Pricing
N14
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 20.4 mPa.s (25 °C)
Expand
View Pricing
D500
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 494 mPa.s (25 °C)
Expand
View Pricing
S200
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 392 mPa.s (25 °C)
Expand
View Pricing
N2
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 2.144 mPa.s (25 °C)
Expand
View Pricing
Page 1 of 2
Page 1 of 2