Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gallic acid monohydrate
Synonym(s):
3,4,5-Trihydroxybenzoic acid monohydrate
Linear Formula:
(HO)
3
C
6
H
2
CO
2
H · H
2
O
CAS Number:
5995-86-8
Molecular Weight:
188.13
EC Number:
205-749-9
Products
Product Category
building blocks (1)
organic acids (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
ACS reagent (1)
Puriss (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
398225
Gallic acid monohydrate,
ACS reagent, ≥98.0%
Expand
View Pricing
27645
Gallic acid monohydrate,
≥99% (HPLC)
Expand
View Pricing