Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gadolinium(III) oxide
Linear Formula:
Gd
2
O
3
CAS Number:
12064-62-9
Molecular Weight:
362.50
EC Number:
235-060-9
Products
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Puriss (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
278513
Gadolinium(III) oxide,
powder, 99.9% trace metals basis
Expand
View Pricing
48220
Gadolinium(III) oxide,
≥99.9%
Expand
View Pricing
203297
Gadolinium(III) oxide,
powder, ≥99.99% trace metals basis
Expand
View Pricing
637335
Gadolinium(III) oxide,
nanopowder, <100 nm particle size (BET), 99.8% trace metals basis
Expand
View Pricing