Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gadolinium
Synonym(s):
Gadolinium
Linear Formula:
Gd
CAS Number:
7440-54-2
Molecular Weight:
157.25
Products
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Quality Segment
Filter Products
Showing 1-20 of 32
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
263087
Gadolinium,
chips
Expand
View Pricing
GF70193369
Gadolinium,
powder, 500 max. part. size (micron), weight 10 g, purity 99.9%
Expand
View Pricing
263060
Gadolinium,
−40 mesh, 99% trace rare earth metals basis
Expand
View Pricing
GF15174390
Gadolinium,
rod, 50mm, diameter 6.35mm, cast, 99%
Expand
View Pricing
GF00594413
Gadolinium,
foil, thickness 0.25 mm, size 100 × 100 mm, purity 99%
Expand
View Pricing
GF28304951
Gadolinium,
lump, 25 mm max. lump size, weight 100 g, purity 99.9%
Expand
View Pricing
GF92536993
Gadolinium,
wire, 1.0 mm diameter, purity 99.9%, length 25 mm
Expand
View Pricing
GF55248453
Gadolinium,
rod, 100mm, diameter 12.5mm, cast, 99%
Expand
View Pricing
GF72371695
Gadolinium,
foil, thickness 0.01 mm, purity 99%, size 25 × 25 mm
Expand
View Pricing
GF96799875
Gadolinium,
rod, 100mm, diameter 6.35mm, cast, 99%
Expand
View Pricing
GF51795232
Gadolinium,
foil, thickness 0.25 mm, size 25 × 25 mm, purity 99%
Expand
View Pricing
GF49402593
Gadolinium,
lump, 25 mm max. lump size, weight 20 g, purity 99.9%
Expand
View Pricing
GF61542323
Gadolinium,
lump, 50 mm max. lump size, weight 100 g, purity 99.9%
Expand
View Pricing
GF98041989
Gadolinium,
rod, 50mm, diameter 12.5mm, cast, 99%
Expand
View Pricing
GF48337487
Gadolinium,
rod, 48mm, diameter 3.0mm, cast, 99%
Expand
View Pricing
GF35943701
Gadolinium,
foil, thickness 0.1 mm, size 25 × 25 mm, purity 99%
Expand
View Pricing
GF68780627
Gadolinium,
foil, thickness 0.05 mm, size 25 × 25 mm, purity 99%
Expand
View Pricing
GF52231711
Gadolinium,
wire reel, 50mm, diameter 0.5mm, hard, 99.9%
Expand
View Pricing
GF92963313
Gadolinium,
foil, thickness 0.1 mm, size 50 × 50 mm, purity 99%
Expand
View Pricing
GF13286369
Gadolinium,
wire reel, 25mm, diameter 0.5mm, hard, 99.9%
Expand
View Pricing
Page 1 of 2
Page 1 of 2