Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Four-way microtube rack
Products
Product Category
racks (7)
Brand
Sigma-Aldrich (6)
Color
Manufacturer
Special Grade
Natural (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
R5651
Four-way microtube rack,
assorted colors
Expand
View Pricing
R5401
Four-way microtube rack,
yellow
Expand
View Pricing
R5276
Four-way microtube rack,
pink
Expand
View Pricing
R5151
Four-way microtube rack,
green
Expand
View Pricing
R5026
Four-way microtube rack,
blue
Expand
View Pricing
R4901
Four-way microtube rack,
natural
Expand
View Pricing
R5526
Four-way microtube rack,
orange
Expand
View Pricing