
Foscarnet Related Compound D

Synonym(s):
Triethyl phosphonoformate, Ethyl diethoxyphospinylformate
Linear Formula:
(C2H5O)2P(O)COOC2H5
CAS Number:
Molecular Weight:
210.16
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2