
Fluroxypyr-1-methylheptyl ester

Synonym(s):
1-Methylheptyl (4-amino-3,5-dichloro-6-fluoro-2-pyridyloxy)acetate, Fluroxypyr meptyl
Empirical Formula (Hill Notation):
C15H21Cl2FN2O3
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing