Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fluorescein isothiocyanate–dextran
Synonym(s):
FITC–Dextran
CAS Number:
60842-46-8
Products
Product Category
carbohydrates (18)
dextrans (12)
Brand
Sigma-Aldrich (18)
Biological Source
Color
Physical Form
Research Area
Tissue
Conjugate
Application
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
46944
Fluorescein isothiocyanate–dextran,
average mol wt 4,000, (FITC:Glucose = 1:250)
Expand
View Pricing
FD4
Fluorescein isothiocyanate–dextran,
average mol wt 3,000-5,000
Expand
View Pricing
46945
Fluorescein isothiocyanate–dextran,
average mol wt 70,000, (FITC:Glucose = 1:250)
Expand
View Pricing
FD10S
Fluorescein isothiocyanate–dextran,
average mol wt 10,000
Expand
View Pricing
FD2000S
Fluorescein isothiocyanate–dextran,
average mol wt 2,000,000
Expand
View Pricing
FD40S
Fluorescein isothiocyanate–dextran,
average mol wt 40,000
Expand
View Pricing
FD40
Fluorescein isothiocyanate–dextran,
average mol wt 40,000
Expand
View Pricing
FD70S
Fluorescein isothiocyanate–dextran,
average mol wt 60,000-76,000
Expand
View Pricing
FD500S
Fluorescein isothiocyanate–dextran,
average mol wt 500,000
Expand
View Pricing
FD20S
Fluorescein isothiocyanate–dextran,
average mol wt 20,000
Expand
View Pricing
FD150S
Fluorescein isothiocyanate–dextran,
average mol wt 150,000
Expand
View Pricing
46946
Fluorescein isothiocyanate–dextran,
average mol wt 150,000, (FITC:Glucose = 1:160)
Expand
View Pricing
90718
Fluorescein isothiocyanate–dextran,
average M
w
70,000
Expand
View Pricing
FD20
Fluorescein isothiocyanate–dextran,
average mol wt 20,000
Expand
View Pricing
FD70
Fluorescein isothiocyanate–dextran,
average M
w
59,000-77,000
Expand
View Pricing
FD250S
Fluorescein isothiocyanate–dextran,
average mol wt 250,000
Expand
View Pricing
52471
Fluorescein isothiocyanate–dextran,
average mol wt 2,000,000
Expand
View Pricing
69658
Fluorescein isothiocyanate–dextran,
average mol wt 150,000
Expand
View Pricing